The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039126.2 | NP_001034215.1 | 2 - 3 | 446 | 66 |
NM_023046.4 | NP_075533.3 | 2 - 3 | 247 | 66 | |
GenBank mRNA | AF155352.1 | - | 2 - 3 | 198 | 66 |
AK006515.1 | - | 2 - 3 | 248 | 66 | |
AK122456.1 | - | 2 - 3 | 302 | 66 | |
AK153774.1 | - | 2 - 3 | 468 | 66 | |
BC034869.1 | - | 2 - 3 | 302 | 66 | |
BC049699.1 | - | 2 - 3 | 247 | 66 | |
BU938650.1 | - | 2 - 3 | 250 | 66 |