The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163189.1 | NP_001156661.1 | 5 - 6 | 1381 | 85 |
GenBank mRNA | BC158112.1 | - | 5 - 6 | 1381 | 85 |