The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048179.1 | NP_001041644.1 | 2 - 3 | 103 | 72 |
NM_001164044.1 | NP_001157516.1 | 2 - 3 | 145 | 72 | |
NM_001164045.1 | NP_001157517.1 | 1 - 2 | 366 | 72 | |
GenBank mRNA | AK005753.1 | - | 2 - 3 | 98 | 72 |
BC099574.1 | - | 1 - 2 | 366 | 72 | |
CA451425.1 | - | 2 - 3 | 103 | 72 |