The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286944.1 | NP_001273873.1 | 1 - 1 | 2547 | 97 |
NM_010592.5 | NP_034722.1 | 1 - 1 | 2547 | 97 | |
GenBank mRNA | BC010572.1 | - | 1 - 1 | 1344 | 97 |
BC023211.1 | - | 1 - 1 | 1338 | 97 | |
BC094068.1 | - | 2 - 2 | 1229 | 97 | |
BQ177858.1 | - | 1 - 1 | 570 | 97 | |
J05205.1 | - | 1 - 1 | 1271 | 97 | |
X15358.1 | - | 1 - 1 | 1259 | 97 |