The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011002.2 | NP_035132.2 | 1 - 2 | 242 | 101 |
GenBank mRNA | DR065535.1 | - | 1 - 2 | 242 | 101 |