Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plin3
Entrez Gene ID
66905
Gene Name
perilipin 3
Gene Aliases
1300012C15Rik, M6prbp1, Tip47
Chromosome Location
Chr.17: 56278962 - 56290511 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025836.3 NP_080112.1 5 - 6 723 83
GenBank mRNA AK004970.1 - 5 - 6 724 83
AK009148.1 - 1 - 2 270 83
AK012517.1 - 5 - 6 724 83
AK049537.1 - 5 - 6 724 83
AK050140.1 - 5 - 6 725 83
AK132313.1 - - 724 83
AK167214.1 - 5 - 6 722 83
BC011116.1 - 5 - 6 719 83

View Full Product Details