Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plin3
Entrez Gene ID
66905
Gene Name
perilipin 3
Gene Aliases
1300012C15Rik, M6prbp1, Tip47
Chromosome Location
Chr.17: 56278962 - 56290511 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025836.3 NP_080112.1 7 - 8 1049 112
GenBank mRNA AK004970.1 - 7 - 8 1050 112
AK009148.1 - 3 - 4 596 112
AK012517.1 - 7 - 8 1050 112
AK049537.1 - 7 - 8 1050 112
AK050140.1 - 7 - 8 1051 112
AK132313.1 - - 1050 112
AK167214.1 - 7 - 8 1048 112
BC011116.1 - 7 - 8 1045 112

View Full Product Details