Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt80
Entrez Gene ID
74127
Gene Name
keratin 80
Gene Aliases
1200016G03Rik, 2310041I20Rik, Kb20
Chromosome Location
Chr.15: 101348315 - 101370125 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028770.2 NP_083046.2 4 - 5 737 84
GenBank mRNA AK004811.2 - 3 - 4 526 84
AK171114.1 - - 626 84
BC119366.1 - 4 - 5 737 84
BC120601.1 - 4 - 5 737 84

View Full Product Details