The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028900.4 | NP_083176.3 | 1 - 2 | 1439 | 66 |
GenBank mRNA | AK016531.1 | - | - | 1388 | 66 |
AK032253.1 | - | 1 - 2 | 1439 | 66 | |
AK181400.1 | - | 1 - 2 | 350 | 66 | |
BC066807.1 | - | 1 - 2 | 1408 | 66 |