Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cldn22
- Entrez Gene ID
-
75677
- Gene Name
- claudin 22
- Gene Aliases
-
2210404A22Rik, 5330431C14Rik, 9530051B05Rik
- Chromosome Location
-
Chr.8: 47824482 - 47825475 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029383.1 |
NP_083659.1 |
1 - 1 |
56 |
89 |
GenBank mRNA |
AK008821.1 |
- |
1 - 1 |
57 |
89 |
- Gene Symbol
- Wwc2
- Entrez Gene ID
-
52357
- Gene Name
- WW, C2 and coiled-coil domain containing 2
- Gene Aliases
-
5330431C14Rik, AU022508, C85457, D8Ertd594e
- Chromosome Location
-
Chr.8: 47827595 - 47990551 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK036263.1 |
- |
- |
|
89 |
AK044079.1 |
- |
- |
|
89 |
View Full Product Details