The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053129.3 | NP_444359.3 | 1 - 1 | 51 | 98 |
GenBank mRNA | AK029277.1 | - | 1 - 1 | 43 | 98 |
AK036169.1 | - | 1 - 1 | 52 | 98 | |
AK143280.1 | - | 1 - 1 | 52 | 98 | |
AK160730.1 | - | 1 - 1 | 52 | 98 |