The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016668.3 | NP_057877.1 | 6 - 7 | 1074 | 89 |
GenBank mRNA | AF033381.1 | - | 6 - 7 | 817 | 89 |
AK077108.1 | - | 6 - 7 | 882 | 89 | |
AK145668.1 | - | 6 - 7 | 916 | 89 | |
AK149457.1 | - | 6 - 7 | 881 | 89 | |
AK166754.1 | - | 6 - 7 | 883 | 89 | |
AK166818.1 | - | 6 - 7 | 882 | 89 | |
BC037004.1 | - | 6 - 7 | 876 | 89 | |
BC093510.1 | - | 6 - 7 | 849 | 89 | |
BC110307.1 | - | 6 - 7 | 1277 | 89 | |
BG078929.2 | - | 4 - 5 | 566 | 89 |