The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243161.1 | NP_001230090.1 | 1 - 2 | 51 | 77 |
NM_183142.4 | NP_898965.1 | 1 - 2 | 51 | 77 | |
NR_040650.1 | 1 - 2 | 51 | 77 | ||
GenBank mRNA | AB288237.1 | - | 1 - 2 | 45 | 77 |
AB288238.1 | - | 1 - 2 | 45 | 77 | |
AK046475.1 | - | 1 - 2 | 53 | 77 | |
AK157746.1 | - | 1 - 2 | 51 | 77 |