Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Serpinb9f
- Entrez Gene ID
-
20709
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade B, member 9f
- Gene Aliases
-
BC052216, NK21, NK21L1, Spi13, ovalbumin
- Chromosome Location
-
Chr.13: 33324077 - 33335370 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_183197.1 |
NP_899020.1 |
3 - 4 |
362 |
103 |
GenBank mRNA |
BC052216.1 |
- |
3 - 4 |
362 |
103 |
- Gene Symbol
- Serpinb9d
- Entrez Gene ID
-
20726
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade B, member 9d
- Gene Aliases
-
AT2, Spi9, ovalbumin
- Chromosome Location
-
Chr.13: 33192959 - 33203980 on Build GRCm38
- Gene Symbol
- Serpinb9g
- Entrez Gene ID
-
93806
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade B, member 9g
- Gene Aliases
-
1600002F03Rik, AW545779, NK21B
- Chromosome Location
-
Chr.13: 33484790 - 33496002 on Build GRCm38
View Full Product Details