Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- Adrm1
- Entrez Gene ID
-
56436
- Gene Name
- adhesion regulating molecule 1
- Gene Aliases
-
1110063P18Rik, 2510006J17Rik, AA408205, ARM-1, AU043535, Arm1, Gp110, Rpn13
- Chromosome Location
-
Chr.2: 180171490 - 180176286 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019822.3 |
NP_062796.2 |
1 - 1 |
52 |
99 |
GenBank mRNA |
BC008974.1 |
- |
1 - 1 |
52 |
99 |
- Gene Symbol
- Gm29886
- Entrez Gene ID
-
102631586
- Gene Name
- predicted gene, 29886
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 180149953 - 180171688 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK131767.1 |
- |
1 - 1 |
|
99 |
View Full Product Details