Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hist2h2ac
- Entrez Gene ID
-
319176
- Gene Name
- histone cluster 2, H2ac
- Gene Aliases
-
H2a-613b
- Chromosome Location
-
Chr.3: 96220413 - 96220880 on Build GRCm38
- Gene Symbol
- Hist2h2aa1
- Entrez Gene ID
-
15267
- Gene Name
- histone cluster 2, H2aa1
- Gene Aliases
-
AI448581, H2a-614, H2a-615, Hist2
- Chromosome Location
-
Chr.3: 96245497 - 96246052 on Build GRCm38
- Gene Symbol
- Hist2h2aa2
- Entrez Gene ID
-
319192
- Gene Name
- histone cluster 2, H2aa2
- Gene Aliases
-
H2a-614, H2a-615
- Chromosome Location
-
Chr.3: 96239779 - 96240374 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178212.3 |
NP_835584.1 |
1 - 1 |
279 |
88 |
View Full Product Details