Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bdnf
Entrez Gene ID
12064
Gene Name
brain derived neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.2: 109674700 - 109727043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048142.1 NP_001041607.1 1 - 1 154 103
GenBank mRNA AK017559.1 - 1 - 1 88 103
AK033127.1 - 1 - 1 100 103
AY057914.1 - 1 - 1 154 103
EF125674.1 - 1 - 1 154 103

View Full Product Details