Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Scarna13
- Entrez Gene ID
-
100306943
- Gene Name
- small Cajal body-specific RNA 1
- Gene Aliases
-
U93
- Chromosome Location
-
Chr.12: 105031075 - 105031349 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028576.1 |
|
1 - 1 |
49 |
68 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE633860.1 |
- |
1 |
184 |
|
Chr. 12 - 105031013 - 105031485 [-] on Build GRCm38 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 3069 |
Mir3069 |
mir-3069, mmu-mir-3069 |
|
Sense |
Chr. - 105031077 - 105031141 on Build GRCm38 |
Downstream |
glutaredoxin 5 homolog (S. cerevisiae) |
Glrx5 |
2310004O13Rik, 2900070E19Rik, AU020725 |
1.3kb
|
Anti-sense |
Chr. - 105032689 - 105040910 on Build GRCm38 |
View Full Product Details