Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mir20a
Entrez Gene ID
387139
Gene Name
microRNA 20a
Gene Aliases
Mir-20, Mirn20, Mirn20a, mir-20a, mmu-mir-20a
Chromosome Location
Chr.14: 115044157 - 115044263 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_029737.1 1 - 1 48 97


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 19a Mir19a Mirn19a, mir-19a, mmu-mir-19a -76.0 Sense Chr. - 115044000 - 115044081 on Build GRCm38
Downstream microRNA 19b-1 Mir19b-1 Mirn19b-1, mir-19b-1 42.0 Sense Chr. - 115044305 - 115044391 on Build GRCm38

View Full Product Details