Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mgat4c
Entrez Gene ID
67569
Gene Name
MGAT4 family, member C
Gene Aliases
9130411I17Rik, GntIVh
Chromosome Location
Chr.10: 101681487 - 102391851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162368.1 NP_001155840.1 4 - 5 470 101
NM_001162369.1 NP_001155841.1 4 - 5 343 101
NM_001205098.1 NP_001192027.1 6 - 7 883 101
NM_026243.4 NP_080519.2 2 - 3 365 101
GenBank mRNA AK018574.1 - 2 - 3 364 101
AK032009.1 - 4 - 5 344 101
AK033482.1 - 2 - 3 366 101
BB611341.1 - 4 - 5 471 101
BC046987.1 - 2 - 3 353 101
CD774691.1 - 6 - 7 651 101

View Full Product Details