The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199270.1 | NP_001186199.1 | 1 - 1 | 1957 | 113 |
GenBank mRNA | AK016845.1 | - | 1 - 1 | 1930 | 113 |