Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H60c
Entrez Gene ID
670558
Gene Name
histocompatibility 60c
Gene Aliases
4632413I24Rik, Gm9491
Chromosome Location
Chr.10: 3256202 - 3268092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204916.2 NP_001191845.1 5 - 6 822 91
GenBank mRNA AB284506.1 - 5 - 6 801 91

View Full Product Details