Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
4921511I17Rik
Entrez Gene ID
70858
Gene Name
RIKEN cDNA 4921511I17 gene
Gene Aliases
-
Chromosome Location
Chr.12: 12351522 - 12392475 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045829.1 4 - 5 382 129
GenBank mRNA AK014875.1 - 4 - 5 383 129


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY714395.1 - 4 - 5 383 Chr. 12 - 12351947 - 12392643 [-] on Build GRCm38 129


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream tubulin, beta 2a, pseudogene 2 Tubb2a-ps2 C79445 -468.6kb Sense Chr. - 11882196 - 11882899 on Build GRCm38
Downstream predicted gene, 34637 Gm34637 313.6kb Sense Chr. - 12667732 - 12669963 on Build GRCm38

View Full Product Details