The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271497.1 | NP_001258426.1 | 6 - 7 | 753 | 86 |
NM_001271498.1 | NP_001258427.1 | 6 - 7 | 847 | 86 | |
NM_001271499.1 | NP_001258428.1 | 6 - 7 | 717 | 86 | |
NM_001271500.1 | NP_001258429.1 | 5 - 6 | 618 | 86 | |
NM_001271501.1 | NP_001258430.1 | 4 - 5 | 525 | 86 | |
NM_008358.2 | NP_032384.1 | 6 - 7 | 816 | 86 | |
NM_133836.2 | NP_598597.1 | 6 - 7 | 869 | 86 | |
NR_073191.1 | 5 - 6 | 671 | 86 | ||
NR_073192.1 | 4 - 5 | 578 | 86 | ||
GenBank mRNA | AK040940.1 | - | 6 - 7 | 699 | 86 |
AK080000.1 | - | 6 - 7 | 699 | 86 | |
AY219715.1 | - | 6 - 7 | 618 | 86 | |
AY219716.1 | - | 5 - 6 | 520 | 86 | |
AY219717.1 | - | 4 - 5 | 427 | 86 | |
AY221616.1 | - | 6 - 7 | 494 | 86 | |
AY221617.1 | - | 5 - 6 | 412 | 86 | |
AY221618.1 | - | 4 - 5 | 319 | 86 | |
AY221619.1 | - | 6 - 7 | 586 | 86 | |
BC022705.1 | - | 6 - 7 | 626 | 86 | |
BC069881.1 | - | 5 - 6 | 577 | 86 | |
BC095982.1 | - | 3 - 4 | 544 | 86 | |
BC132231.1 | - | 5 - 6 | 416 | 86 | |
BC132233.1 | - | 5 - 6 | 416 | 86 | |
BC145348.1 | - | 6 - 7 | 487 | 86 |