The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011877.2 | NP_001011877.1 | 1 - 1 | 318 | 100 |
GenBank mRNA | BC141252.1 | - | 1 - 1 | 318 | 100 |