The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271378.1 | NP_001258307.1 | 2 - 3 | 314 | 227 |
NM_001271379.1 | NP_001258308.1 | 2 - 3 | 314 | 122 | |
NM_011167.3 | NP_035297.1 | 2 - 3 | 311 | 227 | |
NR_073166.1 | 2 - 3 | 236 | 227 | ||
GenBank mRNA | AF090140.1 | - | 2 - 3 | 220 | 227 |
AK005435.1 | - | 2 - 3 | 275 | 227 | |
AK005478.1 | - | 2 - 3 | 315 | 122 | |
BC099469.1 | - | 2 - 3 | 257 | 227 |