Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfam1
Entrez Gene ID
74039
Gene Name
Nfat activating molecule with ITAM motif 1
Gene Aliases
4921501M20Rik, Cnaip
Chromosome Location
Chr.15: 82996736 - 83033397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271411.1 NP_001258340.1 5 - 6 901 113
NM_001271412.1 NP_001258341.1 5 - 6 831 113
NM_001271413.1 NP_001258342.1 5 - 6 740 113
NM_001271414.1 NP_001258343.1 5 - 6 828 113
NM_028728.3 NP_083004.1 5 - 6 758 113
GenBank mRNA AK014802.2 - 5 - 6 759 113
AK048992.1 - 5 - 6 742 113
AK150635.1 - 5 - 6 810 113
AK151187.1 - 5 - 6 810 113
AK151884.1 - 5 - 6 810 113
AK152262.1 - 5 - 6 810 113
AK152499.1 - 5 - 6 810 113
AK152688.1 - 5 - 6 810 113
AK155106.1 - 5 - 6 755 113
AK169918.1 - 5 - 6 755 113
AK170098.1 - 5 - 6 845 113
AK170355.1 - 5 - 6 808 113
AK170442.1 - 5 - 6 810 113
AK170545.1 - 5 - 6 696 113
AK170593.1 - 5 - 6 766 113
AK170811.1 - 5 - 6 766 113
AK170883.1 - 5 - 6 809 113
AK171082.1 - 5 - 6 871 113
AK171086.1 - 5 - 6 779 113
AK171201.1 - 5 - 6 740 113
BC150153.1 - 5 - 6 773 113
BC150488.1 - 5 - 6 773 113

View Full Product Details