Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd8
Entrez Gene ID
100503185
Gene Name
BTB (POZ) domain containing 8
Gene Aliases
EG627196, Gm16115, OTTMUSG00000030610
Chromosome Location
Chr.5: 107437997 - 107491596 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001255991.1 NP_001242920.1 2 - 3 345 128

View Full Product Details