The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_102291.1 | 2 - 3 | 274 | 91 | |
NR_102292.1 | 2 - 3 | 274 | 91 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ring finger protein 207 | Rnf207 | D330010C22Rik, Gm143 | -8.1kb | Anti-sense | Chr. - 152306134 - 152319369 on Build GRCm38 |
Downstream | chromodomain helicase DNA binding protein 5 | Chd5 | 4930532L22Rik, AW060752, B230399N07Rik, CHD-5 | 9.2kb | Sense | Chr. - 152338651 - 152390194 on Build GRCm38 |