Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc17a1
- Entrez Gene ID
-
20504
- Gene Name
- solute carrier family 17 (sodium phosphate), member 1
- Gene Aliases
-
NAPI-1, Napi1, Npt1
- Chromosome Location
-
Chr.13: 23867723 - 23895730 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_121611.1 |
|
8 - 9 |
1648 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 17 (sodium phosphate), member 3 |
Slc17a3 |
AW261723, Npt4 |
-18.0kb
|
Sense |
Chr. - 23836918 - 23860718 on Build GRCm38 |
Downstream |
solute carrier family 17 (sodium phosphate), member 4 |
Slc17a4 |
9130214H05Rik |
17.2kb
|
Anti-sense |
Chr. - 23897879 - 23915009 on Build GRCm38 |
View Full Product Details