Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- E4f1
- Entrez Gene ID
-
13560
- Gene Name
- E4F transcription factor 1
- Gene Aliases
-
p120E4F, phi-AP3
- Chromosome Location
-
Chr.17: 24443778 - 24455590 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CO425601.1 |
- |
3 - 4 |
381 |
|
Chr. 17 - 24445984 - 24447401 [-] on Build GRCm38 |
59 |
GenBankmRNA |
BM947613.1 |
- |
5 - 6 |
563 |
|
Chr. 17 - 24446567 - 24451479 [-] on Build GRCm38 |
59 |
GenBankmRNA |
BQ178864.1 |
- |
3 - 4 |
381 |
|
Chr. 17 - 24446011 - 24447401 [-] on Build GRCm38 |
59 |
GenBankmRNA |
DV648753.1 |
- |
3 - 4 |
356 |
|
Chr. 17 - 24446001 - 24447376 [-] on Build GRCm38 |
59 |
GenBankmRNA |
BG975422.1 |
- |
2 - 3 |
204 |
|
Chr. 17 - 24445465 - 24447137 [-] on Build GRCm38 |
59 |
GenBankmRNA |
CA528677.1 |
- |
3 - 4 |
335 |
|
Chr. 17 - 24446057 - 24447355 [-] on Build GRCm38 |
59 |
GenBankmRNA |
BQ442418.1 |
- |
1 - 2 |
135 |
|
Chr. 17 - 24445174 - 24446914 [-] on Build GRCm38 |
59 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
deoxyribonuclease 1-like 2 |
Dnase1l2 |
4733401H14Rik |
-2.7kb
|
Sense |
Chr. - 24440087 - 24443765 on Build GRCm38 |
Downstream |
RIKEN cDNA 4833447I15 gene |
4833447I15Rik |
|
14.3kb
|
Sense |
Chr. - 24461220 - 24470191 on Build GRCm38 |
View Full Product Details