Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF7
Entrez Gene ID
101106816
Gene Name
ring finger protein 7
Gene Aliases
-
Chromosome Location
Chr.1: 269853117 - 269860121 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003291.4 XP_004003340.1 1 - 2 291 74

View Full Product Details