Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDLIM1
Entrez Gene ID
101107916
Gene Name
PDZ and LIM domain 1
Gene Aliases
-
Chromosome Location
Chr.22: 19253166 - 19291909 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960151.1 XP_027815952.1 6 - 7 916 60

View Full Product Details