Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMAB
Entrez Gene ID
101114057
Gene Name
metabolism of cobalamin associated B
Gene Aliases
-
Chromosome Location
Chr.17: 72831540 - 72842794 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956642.1 XP_027812443.1 3 - 4 337 70

View Full Product Details