Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNB4
Entrez Gene ID
101123499
Gene Name
cholinergic receptor nicotinic beta 4 subunit
Gene Aliases
-
Chromosome Location
Chr.18: 28956801 - 28973512 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017822.4 XP_004017871.2 3 - 4 364 63

View Full Product Details