Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABARAP
Entrez Gene ID
101118285
Gene Name
GABA type A receptor-associated protein
Gene Aliases
-
Chromosome Location
Chr.11: 36397437 - 36400179 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012641.4 XP_004012690.1 3 - 4 1124 94

View Full Product Details