Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBL4A
Entrez Gene ID
101118855
Gene Name
ubiquitin like 4A
Gene Aliases
-
Chromosome Location
Chr.X: 88506033 - 88508759 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963034.1 XP_027818835.1 4 - 5 548 61

View Full Product Details