Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RANGRF
Entrez Gene ID
101107111
Gene Name
RAN guanine nucleotide release factor
Gene Aliases
-
Chromosome Location
Chr.11: 35522000 - 35523440 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012682.4 XP_004012731.1 1 - 2 223 90
XM_012185695.3 XP_012041085.1 1 - 2 224 90

View Full Product Details