Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL5A
Entrez Gene ID
100302313
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
-
Chromosome Location
Chr.2: 169493828 - 169514863 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162555.1 NP_001156027.1 5 - 6 491 63
GenBank mRNA FJ969398.1 - 5 - 6 491 63

View Full Product Details