Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INHA
Entrez Gene ID
101118082
Gene Name
inhibin subunit alpha
Gene Aliases
-
Chromosome Location
Chr.2: 236308613 - 236311576 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308579.1 NP_001295508.1 1 - 2 264 72
GenBank mRNA KP113678.1 - 1 - 2 264 72

View Full Product Details