Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMM17B
Entrez Gene ID
101108454
Gene Name
translocase of inner mitochondrial membrane 17B
Gene Aliases
-
Chromosome Location
Chr.X: 58510068 - 58514074 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963651.1 XP_027819452.1 2 - 3 211 79

View Full Product Details