Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPC24
Entrez Gene ID
101117500
Gene Name
SPC24 component of NDC80 kinetochore complex
Gene Aliases
-
Chromosome Location
Chr.5: 13888576 - 13895236 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969671.1 XP_027825472.1 3 - 4 412 83

View Full Product Details