Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6V1G2
Entrez Gene ID
101123332
Gene Name
ATPase H+ transporting V1 subunit G2
Gene Aliases
-
Chromosome Location
Chr.20: 29580118 - 29582388 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018963.4 XP_004019012.1 2 - 3 390 72

View Full Product Details