Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL36RN
Entrez Gene ID
101111354
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
-
Chromosome Location
Chr.3: 63941854 - 63946453 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966770.1 XP_027822571.1 3 - 4 167 78

View Full Product Details