Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HS3ST5
Entrez Gene ID
101107448
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 5
Gene Aliases
-
Chromosome Location
Chr.8: 26003018 - 26210979 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972353.1 XP_027828154.1 3 - 4 498 81

View Full Product Details