Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL21
Entrez Gene ID
101120765
Gene Name
kelch like family member 21
Gene Aliases
-
Chromosome Location
Chr.12: 50452408 - 50465454 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013754.4 XP_004013803.2 1 - 2 1071 68

View Full Product Details