Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
101122153
Gene Name
ribosome binding factor A
Gene Aliases
-
Chromosome Location
Chr.23: 637480 - 651831 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960705.1 XP_027816506.1 1 - 2 175 77

View Full Product Details