Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB16
Entrez Gene ID
101115385
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
Chromosome Location
Chr.11: 19066285 - 19074412 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098825.2 XP_014954311.2 3 - 4 1378 67

View Full Product Details