Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD11B1
Entrez Gene ID
443409
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.12: 77036851 - 77105871 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009395.2 NP_001009395.1 1 - 2 152 85
XM_012186882.2 XP_012042272.1 2 - 3 375 85
XM_027975568.1 XP_027831369.1 4 - 5 655 85
XM_027975569.1 XP_027831370.1 3 - 4 1015 85
XM_027975570.1 XP_027831371.1 3 - 4 484 85
GenBank mRNA X69561.1 - - 155 85

View Full Product Details