Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC24A5
Entrez Gene ID
100820754
Gene Name
solute carrier family 24 member 5
Gene Aliases
-
Chromosome Location
Chr.7: 64003680 - 64024701 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252181.1 NP_001239110.1 7 - 8 1082 109
GenBank mRNA JF719290.1 - 7 - 8 1082 109

View Full Product Details